3-Bromo-2-fluoro-6-methoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A877648-100MG A877648-250MG A877648-1G
77171-738EA 25 USD
77171-738 77171-740 77595-422
3-Bromo-2-fluoro-6-methoxybenzaldehyde 98%
3-Bromo-2-fluoro-6-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MDL Number: MFCD15528066
CAS Number: 1160653-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR