9-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-9H-purin-6-ol 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A877620-100MG A877620-1MG A877620-1G A877620-5G A877620-25G
77171-734EA 7 USD
77171-734 77171-736 77684-842 77684-843 77893-679
9-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-9H-purin-6-ol 98%, Ambeed.Inc
9-((2R,4S,5R)-5-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-4-hydroxytetrahydrofuran-2-yl)-9H-purin-6-ol

Precaución: For research use only.

Formula: C₃₁H₃₀N₄O₆
MDL Number: MFCD00057879
CAS Number: 93778-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR