N,N-Dimethyl-1,3-phenylenediammonium dichloride 95%

Supplier: Ambeed
Warning

Synonyms: N1,N1-Dimethylbenzene-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-phenylenediamine dihydrochloride, N,N-Dimethyl-m-phenylenediamine dihydrochloride

A877569-100G A877569-1G A877569-25G A877569-5G
77200-118EA 766.32 USD
77200-118 77200-120 77200-122 77200-124
N,N-Dimethyl-1,3-phenylenediammonium dichloride 95%
N,N-Dimethyl-1,3-phenylenediammonium dichloride

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂N₂
MW: 209.12 g/mol
MDL Number: MFCD00012971
CAS Number: 3575-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR