N,N,N',N',N''-Pentamethyldiethylenetriamine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine
A877442-10G
A877442-25G
A877442-100G
77200-116EA
5.15
USD
77200-116
77308-482
77595-420
N,N,N',N',N''-Pentamethyldiethylenetriamine 98%
N,N,N',N',N''-Pentamethyldiethylenetriamine
Precaución: for Research Use Only
Formula:
C₉H₂₃N₃ MW: 173.30 g/mol |
MDL Number:
MFCD00014876 CAS Number: 3030-47-5 UN: 2922 ADR: 8,6.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...