3-Cyclobutyl-3-oxo-propionic acid methyl ester 97%

Supplier: Ambeed
Warning

A877365-1G A877365-5G
77171-730EA 46 USD
77171-730 77171-732
3-Cyclobutyl-3-oxo-propionic acid methyl ester 97%
3-Cyclobutyl-3-oxo-propionic acid methyl ester

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MDL Number: MFCD11974606
CAS Number: 137638-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR