1-(2-Bromoethoxy)-4-methoxybenzene 95%

Supplier: Ambeed
Warning

A875098-1G A875098-250MG A875098-5G
77200-110EA 92.7 USD
77200-110 77200-112 77200-114
1-(2-Bromoethoxy)-4-methoxybenzene 95%
1-(2-Bromoethoxy)-4-methoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
MDL Number: MFCD00017893
CAS Number: 22921-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR