2-tert-Butoxypyridine 95%

Supplier: Ambeed
Warning

Synonyms: 2-(tert-Butoxy)pyridine

A874892-100MG A874892-1G A874892-5G A874892-25G A874892-250MG
77171-720EA 10.3 USD
77171-720 77171-722 77171-724 77573-764 77684-840
2-tert-Butoxypyridine 95%
2-tert-Butoxypyridine

Precaución: For research use only.

Formula: C₉H₁₃NO
MW: 151.21 g/mol
MDL Number: MFCD00234567
CAS Number: 83766-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR