Triphos 98%

Supplier: Ambeed
Warning

Synonyms: (2-((Diphenylphosphino)methyl)-2-methylpropane-1,3-diyl)bis(diphenylphosphine), 1,1,1-Tris(diphenylphosphinomethyl)ethane

A873679-1G A873679-250MG A873679-5G
77200-092EA 56.65 USD
77200-092 77200-094 77200-096
Triphos 98%
Triphos

Precaución: for Research Use Only

Formula: C₄₁H₃₉P₃
MW: 624.684 g/mol
MDL Number: MFCD00014084
CAS Number: 22031-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR