2-Amino-4-isopropylthiazole 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A873189-1G A873189-5G A873189-100MG A873189-250MG
77171-714EA 22 USD
77171-714 77171-716 77684-838 77893-673
2-Amino-4-isopropylthiazole 95%, Ambeed.Inc
2-Amino-4-isopropylthiazole

Precaución: For research use only.

Formula: C₆H₁₀N₂S
MDL Number: MFCD07625985
CAS Number: 79932-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR