6-Bromo-5-(trifluoromethyl)pyridin-3-amine 97%

Supplier: Ambeed
Warning

A869686-100MG A869686-1G A869686-250MG
77171-700EA 81.37 USD
77171-700 77171-702 77171-704
6-Bromo-5-(trifluoromethyl)pyridin-3-amine 97%
6-Bromo-5-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₄BrF₃N₂
MW: 241.009 g/mol
MDL Number: MFCD26959067
CAS Number: 1642844-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR