5-Fluoro-1H-indazole-3-carbaldehyde 98%

Supplier: Ambeed
Warning

A868419-1G A868419-250MG A868419-5G A868419-100MG
77171-692EA 127 USD
77171-692 77171-694 77171-696 77893-669
5-Fluoro-1H-indazole-3-carbaldehyde 98%
5-Fluoro-1H-indazole-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅FN₂O
MDL Number: MFCD06739142
CAS Number: 485841-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR