1-(3-Methoxyphenyl)ethanol 97%

Supplier: Ambeed
Warning

A868022-250MG A868022-1G A868022-5G
77200-074EA 11 USD
77200-074 77747-637 77747-638
1-(3-Methoxyphenyl)ethanol 97%
1-(3-Methoxyphenyl)ethanol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00021866
CAS Number: 23308-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR