5-Bromo-2-propyl-1H-imidazole 97%

Supplier: Ambeed
Warning

A867596-100MG A867596-1G A867596-250MG A867596-5G
77200-066EA 110.21 USD
77200-066 77200-068 77200-070 77200-072
5-Bromo-2-propyl-1H-imidazole 97%
5-Bromo-2-propyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₆H₉BrN₂
MW: 189.06 g/mol
MDL Number: MFCD19103374
CAS Number: 1268334-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR