2-Amino-5-chloro-3-iodobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A867559-1G
A867559-250MG
A867559-5G
77200-060EA
12
USD
77200-060
77200-062
77200-064
2-Amino-5-chloro-3-iodobenzoic acid 97%
2-Amino-5-chloro-3-iodobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₅ClINO₂ |
MDL Number:
MFCD07780627 CAS Number: 64724-23-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...