Methyl 6-methoxy-1H-indole-3-carboxylate 96%

Supplier: Ambeed
Warning

A867098-1G A867098-250MG A867098-5G
77171-686EA 76.22 USD
77171-686 77171-688 77171-690
Methyl 6-methoxy-1H-indole-3-carboxylate 96%
Methyl 6-methoxy-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₃
MW: 205.20 g/mol
MDL Number: MFCD09836008
CAS Number: 131424-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR