Benzyl 3-methylpiperazine-1-carboxylate 95%

Supplier: Ambeed
Warning

A860199-1G A860199-250MG A860199-5G
77171-680EA 53 USD
77171-680 77171-682 77171-684
Benzyl 3-methylpiperazine-1-carboxylate 95%
Benzyl 3-methylpiperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD07367776
CAS Number: 84477-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR