1,2-Bis(phenylthio)ethane 98%

Supplier: Ambeed
Warning

A859544-100G A859544-25G A859544-5G
77199-982EA 111.24 USD
77199-982 77199-984 77199-986
1,2-Bis(phenylthio)ethane 98%
1,2-Bis(phenylthio)ethane

Precaución: for Research Use Only

Formula: C₁₄H₁₄S₂
MDL Number: MFCD00014085
CAS Number: 622-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR