5-Ethylisoxazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A857833-1G A857833-250MG A857833-5G
77171-678EA 93 USD
77171-678 77614-530 77893-662
5-Ethylisoxazole-3-carboxylic acid 96%
5-Ethylisoxazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MDL Number: MFCD05864544
CAS Number: 52320-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR