1-(3,4-Dimethoxyphenyl)propan-1-one 95%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Dimethoxyphenyl)propan-1-one

A856830-1G A856830-250MG A856830-5G
77199-966EA 70.04 USD
77199-966 77199-968 77199-970
1-(3,4-Dimethoxyphenyl)propan-1-one 95%
1-(3,4-Dimethoxyphenyl)propan-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.22 g/mol
MDL Number: MFCD00482089
CAS Number: 1835-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR