Ethyl 2-(4-chloro-2-fluorophenyl)acetate 95%

Supplier: Ambeed
Warning

A854323-250MG A854323-5G
77171-656EA 66 USD
77171-656 77171-658
Ethyl 2-(4-chloro-2-fluorophenyl)acetate 95%
Ethyl 2-(4-chloro-2-fluorophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClFO₂
MDL Number: MFCD01570543
CAS Number: 188424-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR