3-Fluoro-4-methylcinnamic acid 97%

Supplier: Ambeed
Warning

A853759-1G A853759-250MG A853759-5G
77199-954EA 30 USD
77199-954 77199-956 77199-958
3-Fluoro-4-methylcinnamic acid 97%
3-Fluoro-4-methylcinnamic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₂
MDL Number: MFCD01631413
CAS Number: 261951-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR