(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis((S)-(2-methoxyphenyl)(phenyl)phosphino)ethane, (S,S)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], (S,S)-(+)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane, (+)-DIPAMP, (S,S)-DIPAMP

A850656-250MG A850656-100MG
77199-952EA 181.28 USD
77199-952 77747-618
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 98%
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane

Precaución: for Research Use Only

Formula: C₂₈H₂₈O₂P₂
MW: 458.48 g/mol
MDL Number: MFCD05863547
CAS Number: 97858-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR