3-Chloro-4-methylphenylhydrazine hydrochloride 97%

Supplier: Ambeed
Warning

A850119-100G A850119-25G A850119-5G
77199-946EA 91 USD
77199-946 77199-948 77199-950
3-Chloro-4-methylphenylhydrazine hydrochloride 97%
3-Chloro-4-methylphenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂·HCl
MDL Number: MFCD00012936
CAS Number: 54812-56-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR