10-Undecen-1-ol 98%
Supplier: Ambeed
|
Warning
|
A849391-100G
A849391-1G
A849391-25G
A849391-5G
A849391-500G
77199-938EA
41
USD
77199-938
77199-940
77199-942
77199-944
77747-616
10-Undecen-1-ol 98%
10-Undecen-1-ol
Precaución: for Research Use Only
Formula:
C₁₁H₂₂O |
MDL Number:
MFCD00004750 CAS Number: 112-43-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...