Dimethylphenylvinylsilane 98% stabilized

Supplier: Ambeed
Danger

Synonyms: Dimethyl(phenyl)(vinyl)silane, Vinylphenyldimethylsilane

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A848051-1G A848051-25G A848051-5G
77171-642EA 7.21 USD
77171-642 77171-644 77171-646
Dimethylphenylvinylsilane 98% stabilized
Dimethylphenylvinylsilane

Precaución: for Research Use Only

Formula: C₁₀H₁₄Si
MW: 162.31 g/mol
MDL Number: MFCD00053857
CAS Number: 1125-26-4
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR