G-749 98+%

Supplier: Ambeed
Warning

Synonyms: G-749, 8-Bromo-2-((1-methylpiperidin-4-yl)amino)-4-((4-phenoxyphenyl)amino)pyrido[4,3-d]pyrimidin-5(6H)-one

A845930-10MG A845930-1MG A845930-25MG A845930-5MG
77171-634EA 63.86 USD
77171-634 77171-636 77171-638 77171-640
G-749 98+%
Denfivontinib

Precaución: for Research Use Only

Formula: C₂₅H₂₅BrN₆O₂
MW: 521.41 g/mol
MDL Number: MFCD28167815
CAS Number: 1457983-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR