1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane 98%, Ambeed.Inc

Supplier: Ambeed
Danger

A844649-25G A844649-100G A844649-500G
77171-632EA 16 USD
77171-632 77684-809 77893-649
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane 98%, Ambeed.Inc
1,1,5,5-Tetramethyl-3,3-diphenyltrisiloxane

Precaución: For research use only.

Formula: C₁₆H₂₄O₂Si₃
MDL Number: MFCD00411646
CAS Number: 17875-55-7
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR