4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 98%
Supplier: Ambeed
|
Warning
|
A844373-1G
A844373-250MG
A844373-5G
77171-626EA
18
USD
77171-626
77171-628
77171-630
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane 98%
4,4,5,5-Tetramethyl-2-[3-(triphenylen-2-yl)phenyl]-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₃₀H₂₇BO₂ |
MDL Number:
MFCD28130383 CAS Number: 1115639-92-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...