(S)-(-)-2-Amino-3-benzyloxy-1-propanol 98%

Supplier: Ambeed
Danger

Synonyms: H-D-Ser(Bzl)-ol

A841513-1G A841513-5G
77199-932EA 147.29 USD
77199-932 77199-934
(S)-(-)-2-Amino-3-benzyloxy-1-propanol 98%
(S)-(-)-2-Amino-3-benzyloxy-1-propanol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MW: 181.23 g/mol
MDL Number: MFCD04112477
CAS Number: 58577-88-1
UN: 3267
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR