Methyl 3-nitro-1-naphthoate 97%

Supplier: Ambeed
Warning

A840107-100MG A840107-250MG
77171-610EA 77.25 USD
77171-610 77171-612
Methyl 3-nitro-1-naphthoate 97%
Methyl 3-nitro-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉NO₄
MW: 231.20 g/mol
MDL Number: MFCD00085480
CAS Number: 13772-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR