1,3-Dicyclohexylimidazolium tetrafluoroborate 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dicyclohexylimidazolium tetrafluoroborate salt

A837582-1G A837582-250MG A837582-5G A837582-25G
77199-914EA 39.14 USD
77199-914 77199-916 77552-564 77747-596
1,3-Dicyclohexylimidazolium tetrafluoroborate 98%
1,3-Dicyclohexylimidazolium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₁₅H₂₅BF₄N₂
MW: 320.18 g/mol
MDL Number: MFCD04973310
CAS Number: 286014-38-8
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR