(S)-2-(1-(tert-Butoxycarbonyl)piperidin-2-yl)acetic acid 96%

Supplier: Ambeed
Warning

A837461-100MG A837461-250MG A837461-1G
77199-910EA 62.83 USD
77199-910 77199-912 77618-338
(S)-2-(1-(tert-Butoxycarbonyl)piperidin-2-yl)acetic acid 96%
(S)-2-(1-(tert-Butoxycarbonyl)piperidin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₁NO₄
MW: 243.3 g/mol
MDL Number: MFCD06656439
CAS Number: 159898-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR