1-(3-Bromophenyl)-1H-pyrazole 98%

Supplier: Ambeed
Warning

A837274-1G A837274-25G A837274-5G
77171-590EA 17.51 USD
77171-590 77171-592 77171-594
1-(3-Bromophenyl)-1H-pyrazole 98%
1-(3-Bromophenyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
MDL Number: MFCD06797472
CAS Number: 294877-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR