(R)-(+)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane 98%
Supplier: Ambeed
|
Warning
|
A837083-100MG
A837083-1G
A837083-50MG
A837083-250MG
77171-584EA
235
USD
77171-584
77171-586
77171-588
77614-502
(R)-(+)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane 98%
(R)-(+)-7,7'-Bis[di(3,5-dimethylphenyl)phosphino]-2,2',3,3'-tetrahydro-1,1'-spirobiindane
Precaución: for Research Use Only
Formula:
C₄₉H₅₀P₂ |
MDL Number:
MFCD08459343 CAS Number: 917377-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...