1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluoropiperidin-1-yl)azetane dihydrochloride, 3-(4-Fluoropiperidin-1-yl)azetidine dihydrochloride

A836660-100MG A836660-1G A836660-250MG
77171-578EA 60.77 USD
77171-578 77171-580 77171-582
1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride 98+%
1-(Azetidin-3-yl)-4-fluoropiperidine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₅FN₂·2ClH
MW: 231.138 g/mol
MDL Number: MFCD11867903
CAS Number: 194427-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR