(1-Methyl-1H-benzo[d]imidazol-5-yl)boronic acid 98+%

Supplier: Ambeed
Danger

A836533-250MG A836533-5G A836533-1G
77171-574EA 70 USD
77171-574 77171-576 77893-628
(1-Methyl-1H-benzo[d]imidazol-5-yl)boronic acid 98+%
(1-Methyl-1H-benzo[d]imidazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD16036273
CAS Number: 1107627-21-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR