(2-Acetamidophenyl)boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamidobenzeneboronic acid

A836327-1G A836327-250MG A836327-5G
77199-904EA 86 USD
77199-904 77199-906 77199-908
(2-Acetamidophenyl)boronic acid 98%
(2-Acetamidophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BNO₃
MW: 178.98 g/mol
MDL Number: MFCD02179474
CAS Number: 169760-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR