7-Bromo-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one 96%
Supplier: Ambeed
|
Warning
|
A835303-100MG
A835303-1G
A835303-250MG
77171-568EA
44
USD
77171-568
77171-570
77171-572
7-Bromo-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one 96%
7-Bromo-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one
Precaución: for Research Use Only
Formula:
C₁₀H₁₀BrNO |
MDL Number:
MFCD02168781 CAS Number: 3951-89-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...