(4-Nitro-1H-pyrazol-1-yl)acetonitrile 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Nitro-1H-pyrazole-1-acetonitrile
A834573-100MG
A834573-1G
A834573-250MG
A834573-5G
77199-888EA
27.81
USD
77199-888
77199-890
77199-892
77199-894
(4-Nitro-1H-pyrazol-1-yl)acetonitrile 97%
(4-Nitro-1H-pyrazol-1-yl)acetonitrile
Precaución: for Research Use Only
Formula:
C₅H₄N₄O₂ MW: 152.11 g/mol |
MDL Number:
MFCD04969696 CAS Number: 1001500-47-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...