Ethyl-2-methylquinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methylquinoline-3-carboxylate, 2-Methyl-quinoline-3-carboxylic acid ethyl ester

A831805-1G A831805-250MG A831805-5G
77171-562EA 95.79 USD
77171-562 77171-564 77171-566
Ethyl-2-methylquinoline-3-carboxylate 97%
Ethyl-2-methylquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₂
MW: 215.25 g/mol
MDL Number: MFCD00195614
CAS Number: 15785-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR