4-(Heptyloxy)benzonitrile 95+%

Supplier: Ambeed
Warning

A831736-1G A831736-250MG
77199-882EA 79 USD
77199-882 77199-884
4-(Heptyloxy)benzonitrile 95+%
4-(Heptyloxy)benzonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO
MW: 217.30 g/mol
MDL Number: MFCD00143323
CAS Number: 29147-88-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR