1-Boc-3-(methylamino)piperidine 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine

A831446-10G A831446-1G A831446-250MG A831446-5G
77171-556EA 109.18 USD
77171-556 77171-558 77171-560 77684-791
1-Boc-3-(methylamino)piperidine 97%
1-Boc-3-(methylamino)piperidine

Precaución: For research use only.

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD04114980
CAS Number: 392331-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR