4-Bromoisoxazol-3-amine 97%

Supplier: Ambeed
Warning

A831351-250MG A831351-5G
77199-880EA 112.27 USD
77199-880 77747-592
4-Bromoisoxazol-3-amine 97%
4-Bromoisoxazol-3-amine

Precaución: for Research Use Only

Formula: C₃H₃BrN₂O
MW: 162.97 g/mol
MDL Number: MFCD21952501
CAS Number: 1519318-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR