3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 97%

Supplier: Ambeed
Warning

Synonyms: di-tBu-Mes-Acr+BF4-, 3,6-Di-tert-butyl-9-mesityl-10-phenylacridin-10-ium tetrafluoroborate

A830612-100MG A830612-250MG A830612-5G A830612-1G
77199-866EA 49.44 USD
77199-866 77199-868 77199-870 77552-548
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 97%
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₃₆H₄₀N·BF₄
MW: 573.514 g/mol
MDL Number: MFCD31381908
CAS Number: 1810004-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR