5,6-Dimethoxy-1H-indazole 98%

Supplier: Ambeed
Warning

A829674-100MG A829674-1G A829674-250MG A829674-5G
77171-544EA 56.65 USD
77171-544 77171-546 77171-548 77171-550
5,6-Dimethoxy-1H-indazole 98%
5,6-Dimethoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₂
MW: 178.19 g/mol
MDL Number: MFCD07371563
CAS Number: 7746-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR