6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%

Supplier: Ambeed
Warning

A828475-10G A828475-1G A828475-25G A828475-5G
77171-536EA 54 USD
77171-536 77171-538 77171-540 77171-542
6-Nitro-2H-1,4-benzoxazin-3(4H)-one 98%
6-Nitro-2H-1,4-benzoxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₆N₂O₄
MDL Number: MFCD03425744
CAS Number: 81721-87-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR