3,5-Di-tert-butyl-2-methoxyaniline 97%

Supplier: Ambeed
Warning

A826614-100MG A826614-1G A826614-250MG A826614-5G
77199-832EA 44 USD
77199-832 77199-834 77199-836 77199-838
3,5-Di-tert-butyl-2-methoxyaniline 97%
3,5-Di-tert-butyl-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₁₅H₂₅NO
MDL Number: MFCD12197084
CAS Number: 893397-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR