Fmoc-Sec(mob)-OH 98%

Supplier: Ambeed
Danger

Synonyms: Fmoc-Sec(Mob)-OH

A826509-100MG A826509-5G A826509-1G
77171-532EA 101 USD
77171-532 77171-534 77552-528
Fmoc-Sec(mob)-OH 98%
Fmoc-Sec(mob)-OH

Precaución: for Research Use Only

Formula: C₂₆H₂₅NO₅Se
MDL Number: MFCD03788170
CAS Number: 150308-80-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR