1-Acetyl-2-pyrrolidone 97%

Supplier: Ambeed
Warning

Synonyms: 1-Acetylpyrrolidin-2-one

A826300-100G A826300-1G A826300-25G A826300-5G
77199-824EA 238.96 USD
77199-824 77199-826 77199-828 77199-830
1-Acetyl-2-pyrrolidone 97%
1-Acetyl-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₆H₉NO₂
MW: 127.14 g/mol
MDL Number: MFCD00014101
CAS Number: 932-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR