Ethyl ((1R,3aR,4aR,6R,8aR,9S,9aS)-9-((E)-2-(5-(3-fluorophenyl)pyridin-2-yl)vinyl)-1-methyl-3-oxododecahydronaphtho[2,3-c]furan-6-yl)carbamate ≥99%

Supplier: Ambeed
Warning

Synonyms: Vorapaxar

A824083-10MG A824083-1MG A824083-25MG A824083-50MG A824083-5MG
77171-516EA 92.7 USD
77171-516 77171-518 77171-520 77171-522 77171-524
Ethyl ((1R,3aR,4aR,6R,8aR,9S,9aS)-9-((E)-2-(5-(3-fluorophenyl)pyridin-2-yl)vinyl)-1-methyl-3-oxododecahydronaphtho[2,3-c]furan-6-yl)carbamate ≥99%
Ethyl ((1R,3aR,4aR,6R,8aR,9S,9aS)-9-((E)-2-(5-(3-fluorophenyl)pyridin-2-yl)vinyl)-1-methyl-3-oxododecahydronaphtho[2,3-c]furan-6-yl)carbamate

Precaución: for Research Use Only

Formula: C₂₉H₃₃FN₂O₄
MW: 492.58 g/mol
MDL Number: MFCD16038876
CAS Number: 618385-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR